Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017550.1 | NP_060020.1 | 3, 4 | 259 | ||
XM_005259582.4 | XP_005259639.1 | 3, 4 | 843 | |||
XM_006722769.3 | XP_006722832.1 | 3, 4 | 847 | |||
XM_011528072.2 | XP_011526374.1 | 3, 4 | 845 | |||
XM_011528073.1 | XP_011526375.1 | 3, 4 | 261 | |||
XM_011528074.2 | XP_011526376.1 | 3, 4 | 849 | |||
XM_011528075.2 | XP_011526377.1 | 3, 4 | 1366 | |||
XM_011528077.2 | XP_011526379.1 | 4, 5 | 3207 | |||
GenBank mRNA | AB033019.1 | - | 247 | |||
BC028203.1 | - | 3, 4 | 259 | 5201526 |