Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GON4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282856.1 | NP_001269785.1 | 2 | 208 | ||
NM_001282858.1 | NP_001269787.1 | 2 | 260 | |||
NM_001282860.1 | NP_001269789.1 | 2 | 260 | |||
NM_001282861.1 | NP_001269790.1 | 2 | 260 | |||
NM_032292.5 | NP_115668.4 | 2 | 248 | |||
XM_005245284.3 | XP_005245341.1 | 1 | 114 | |||
XM_006711393.3 | XP_006711456.1 | 2 | 219 | |||
XM_006711394.3 | XP_006711457.1 | 2 | 205 | |||
XM_011509658.2 | XP_011507960.1 | 2 | 187 | |||
XM_011509659.2 | XP_011507961.1 | 1 | 105 | |||
GenBank mRNA | AB195687.1 | - | 2 | 260 | ||
AB195688.1 | - | 2 | 248 | |||
AB232668.1 | - | 2 | 136 | |||
AK098734.1 | - | 2 | 179 | |||
AY335490.2 | - | 2 | 163 | |||
AY335491.2 | - | 2 | 208 | |||
AY335492.2 | - | 2 | 134 | |||
BC114629.1 | - | 2 | 135 | 40082464 | ||
BC115703.1 | - | 2 | 147 | 40082468 | ||
BX537764.1 | - | 2 | 136 | |||
BX648043.1 | - | 2 | 159 | |||
BX648802.1 | - | 2 | 183 |