Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCAF16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017741.3 | NP_060211.3 | 3 | 1325 | ||
XM_005248169.3 | XP_005248226.1 | 3 | 1313 | |||
XM_005248171.3 | XP_005248228.1 | 2 | 1209 | |||
XM_006713966.3 | XP_006714029.1 | 4 | 1484 | |||
XM_017008327.1 | XP_016863816.1 | 2 | 2778 | |||
XM_017008328.1 | XP_016863817.1 | 2 | 2763 | |||
GenBank mRNA | AK000287.1 | - | 1 | 447 | ||
AK056116.1 | - | 3 | 1325 | |||
BC050697.1 | - | 2 | 986 | 6044417 | ||
BC068025.1 | - | 1 | 624 | 4593954 | ||
BC101716.1 | - | 1 | 335 | 8069222 | ||
BC101718.1 | - | 1 | 335 | 8069224 |