Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF26B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018012.3 | NP_060482.2 | 2 | 560 | ||
GenBank mRNA | AK125187.1 | - | 2 | 561 | ||
AK293964.1 | - | 2 | 475 | |||
AK297303.1 | - | 2 | 556 | |||
AY923834.1 | - | 2 | 561 |