Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SRBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018079.4 | NP_060549.4 | 11 | 1518 | ||
XM_011532946.1 | XP_011531248.1 | 11 | 1519 | |||
XM_017004417.1 | XP_016859906.1 | 11 | 1519 | |||
XM_017004418.1 | XP_016859907.1 | 3 | 1120 | |||
GenBank mRNA | AK001241.1 | - | 5 | 415 | ||
AK056536.1 | - | 11 | 1488 | |||
AK301122.1 | - | 3 | 217 | |||
AK316222.1 | - | 3 | 198 | |||
BC032538.1 | - | 5 | 493 | 5528417 |