Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009552.1 | NP_001009552.1 | 3, 4 | 892 | ||
GenBank mRNA | AF444006.1 | - | 3, 4 | 478 | ||
AK091875.1 | - | 4, 5 | 656 | |||
BC012022.1 | - | 3, 4 | 503 | 4454972 | ||
BT019526.1 | - | 3, 4 | 477 | |||
CR541747.1 | - | 3, 4 | 477 | |||
J03805.1 | - | 3, 4 | 475 | |||
X12656.1 | - | 3, 4 | 498 |