Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018151.4 | NP_060621.3 | 32 | 6953 | ||
XM_005246665.3 | XP_005246722.1 | 32 | 6833 | |||
XM_011511395.2 | XP_011509697.1 | 31 | 4089 | |||
XM_017004422.1 | XP_016859911.1 | 32 | 6902 | |||
XM_017004423.1 | XP_016859912.1 | 31 | 6740 | |||
XM_017004424.1 | XP_016859913.1 | 31 | 6770 | |||
XM_017004425.1 | XP_016859914.1 | 30 | 7078 | |||
GenBank mRNA | AK000323.1 | - | 3 | 1128 | ||
AK074349.1 | - | 2 | 146 | |||
AL080129.1 | - | 3 | 1613 | |||
AY584066.1 | - | 31 | 6792 | |||
AY585745.1 | - | 32 | 6902 | |||
AY727913.1 | - | 32 | 6953 |