Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193360.1 | NP_001180289.1 | 6 | 1193 | ||
NM_001193361.1 | NP_001180290.1 | 5 | 1109 | |||
NM_001193362.1 | NP_001180291.1 | 6 | 1352 | |||
NM_001193363.1 | NP_001180292.1 | 5 | 1268 | |||
NM_018199.3 | NP_060669.1 | 4 | 729 | |||
NR_034164.1 | 7 | 1283 | ||||
NR_034165.1 | 7 | 1442 | ||||
XM_005267817.3 | XP_005267874.1 | 4 | 659 | |||
XM_011536908.2 | XP_011535210.1 | 4 | 747 | |||
XM_017021421.1 | XP_016876910.1 | 5 | 792 | |||
GenBank mRNA | AK001600.1 | - | 4 | 657 | ||
AK056025.1 | - | 7 | 1229 | |||
AK292563.1 | - | 6 | 1352 | |||
AK295601.1 | - | 5 | 1228 | |||
AL834434.1 | - | 4 | 651 | |||
BC001962.1 | - | 4 | 789 | 3537432 | ||
BX647767.1 | - | 5 | 1059 | |||
CR936728.1 | - | 6 | 1068 | |||
CR936729.1 | - | 5 | 1076 |