Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAVER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018211.3 | NP_060681.2 | 9 | 1554 | ||
XM_006710738.3 | XP_006710801.2 | 9 | 1700 | |||
GenBank mRNA | AB046799.1 | - | 9 | 1554 | ||
AL359613.1 | - | 2 | 160 | |||
BC065303.1 | - | 7 | 984 |