Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161575.1 | NP_001155047.1 | 9 | 1664 | ||
NM_018296.5 | NP_060766.5 | 12 | 1934 | |||
XM_005256025.2 | XP_005256082.1 | 12 | 1934 | |||
XM_005256026.2 | XP_005256083.1 | 11 | 1493 | |||
XM_005256027.2 | XP_005256084.1 | 11 | 1788 | |||
XM_005256028.1 | XP_005256085.1 | 11 | 1806 | |||
XM_011523200.1 | XP_011521502.1 | 11 | 1788 | |||
XM_011523201.1 | XP_011521503.1 | 9 | 1554 | |||
XM_011523202.2 | XP_011521504.1 | 8 | 1409 | |||
XM_011523203.1 | XP_011521505.1 | 9 | 1316 | |||
XM_011523204.1 | XP_011521506.1 | 10 | 1733 | |||
XM_011523205.1 | XP_011521507.1 | 11 | 1824 | |||
XM_017023400.1 | XP_016878889.1 | 12 | 1934 | |||
XM_017023401.1 | XP_016878890.1 | 9 | 1037 | |||
XM_017023402.1 | XP_016878891.1 | 7 | 860 | |||
GenBank mRNA | AK001866.1 | - | 8 | 1567 | ||
AK292311.1 | - | 9 | 1643 | |||
AK297329.1 | - | 6 | 1001 | |||
AK301766.1 | - | 7 | 1306 | |||
AY313780.1 | - | 9 | 1627 | |||
AY963267.1 | - | 12 | 1934 | |||
BC018031.1 | - | 11 | 1428 | |||
BC026156.2 | - | 12 | 1934 | 4828487 |