Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC29A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174098.1 | NP_001167569.1 | 2 | 95 | ||
NM_018344.5 | NP_060814.4 | 2 | 95 | |||
NR_033413.1 | 2 | 95 | ||||
NR_033414.1 | 2 | 95 | ||||
XM_006717910.3 | XP_006717973.1 | 2 | 487 | |||
XM_017016377.1 | XP_016871866.1 | 2 | 392 | |||
XM_017016378.1 | XP_016871867.1 | 2 | 395 | |||
GenBank mRNA | AF326987.1 | - | 1 | 43 | ||
AK314497.1 | - | 2 | 63 | |||
AK316152.1 | - | 2 | 80 | |||
AX775915.1 | - | 2 | 95 | |||
AX775917.1 | - | 2 | 95 | |||
AX775919.1 | - | 2 | 95 | |||
AX775921.1 | - | 2 | 95 | |||
AY288928.1 | - | 2 | 55 | |||
BC120996.1 | - | 2 | 73 | 40116513 | ||
BC120997.1 | - | 2 | 73 | 40116514 | ||
DC319920.1 | - | 2 | 95 |