Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MNS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018365.2 | NP_060835.1 | 2 | 329 | ||
GenBank mRNA | AK002084.1 | - | 2 | 329 | ||
AK057542.1 | - | 1 | 143 | |||
BC031046.1 | - | 2 | 242 | 5273227 | ||
BC034991.2 | - | 2 | 296 | 4823345 |