Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM63B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318792.1 | NP_001305721.1 | 16 | 1672 | ||
NM_018426.2 | NP_060896.1 | 16 | 1638 | |||
XM_005249211.3 | XP_005249268.1 | 18 | 2018 | |||
XM_005249213.4 | XP_005249270.1 | 17 | 1945 | |||
XM_005249214.4 | XP_005249271.1 | 15 | 1532 | |||
XM_005249215.4 | XP_005249272.1 | 16 | 1837 | |||
XM_005249217.1 | XP_005249274.1 | 16 | 1710 | |||
XM_006715135.1 | XP_006715198.1 | 16 | 1694 | |||
XM_017010999.1 | XP_016866488.1 | 18 | 1895 | |||
XM_017011000.1 | XP_016866489.1 | 17 | 1876 | |||
XM_017011001.1 | XP_016866490.1 | 15 | 1576 | |||
XM_017011002.1 | XP_016866491.1 | 15 | 1679 | |||
GenBank mRNA | AL353957.1 | - | 2 | 96 | ||
AL834238.1 | - | 9 | 936 | |||
BC022095.1 | - | 13 | 1275 | |||
BC062989.1 | - | 4 | 464 | |||
BC136769.1 | - | 15 | 1514 | 9020759 |