Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114118.2 | NP_001107590.1 | 13 | 1783 | ||
XM_005256717.2 | XP_005256774.1 | 13 | 1783 | |||
XM_006721556.2 | XP_006721619.1 | 12 | 1672 | |||
XM_011523954.2 | XP_011522256.1 | 13 | 1700 | |||
XM_011523955.2 | XP_011522257.1 | 12 | 1554 | |||
XM_017024826.1 | XP_016880315.1 | 12 | 1670 | |||
XM_017024827.1 | XP_016880316.1 | 7 | 1257 | |||
XM_017024828.1 | XP_016880317.1 | 8 | 1075 | |||
XM_017024829.1 | XP_016880318.1 | 8 | 1075 | |||
GenBank mRNA | AJ277841.1 | - | 8 | 1080 | ||
AK125048.1 | - | 10 | 2472 | |||
AK223470.1 | - | 6 | 959 | |||
BC010707.1 | - | 6 | 898 |