Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037639.2 | NP_001032728.1 | 7 | 795 | ||
NM_018622.6 | NP_061092.3 | 8 | 945 | |||
NR_136893.1 | 8 | 1001 | ||||
XM_005247582.4 | XP_005247639.1 | 8 | 940 | |||
XM_017006801.1 | XP_016862290.1 | 7 | 788 | |||
XM_017006803.1 | XP_016862292.1 | 7 | 842 | |||
GenBank mRNA | AF197937.1 | - | 8 | 883 | ||
AK024765.1 | - | 8 | 917 | |||
BC003653.1 | - | 7 | 782 | 3537438 | ||
BC014058.2 | - | 8 | 903 | 3937238 | ||
CA417847.1 | - | 3 | 478 |
Gene Symbol: AIG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017010927.1 | XP_016866416.1 | 6 | 1314 | ||
XM_017010928.1 | XP_016866417.1 | 5 | 1180 |