Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300752.1 | NP_001287681.1 | 8 | 2053 | ||
NM_001300759.1 | NP_001287688.1 | 9 | 2072 | |||
NM_018700.3 | NP_061170.2 | 9 | 2288 | |||
XM_017009621.1 | XP_016865110.1 | 11 | 2300 | |||
XM_017009622.1 | XP_016865111.1 | 11 | 3017 | |||
GenBank mRNA | AB196558.1 | - | 8 | 1544 | ||
AJ272269.1 | - | 9 | 1905 | |||
AK058179.1 | - | 4 | 929 | |||
AK296389.1 | - | 9 | 2070 | |||
AK316422.1 | - | 9 | 1975 | |||
BC130334.1 | - | 9 | 1957 | 40146365 |