Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2Q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017582.6 | NP_060052.3 | 8 | 1033 | ||
GenBank mRNA | AJ243666.1 | - | 2 | 100 | ||
AK056388.1 | - | 4 | 333 | |||
AK293986.1 | - | 8 | 656 | |||
AY112698.1 | - | 8 | 1008 | |||
AY948200.1 | - | 8 | 1016 | |||
BC000848.1 | - | 4 | 367 | |||
BC009286.2 | - | 3 | 171 | |||
BC061583.1 | - | 4 | 345 | |||
BC070158.1 | - | 7 | 558 | |||
BC087836.1 | - | 6 | 444 | |||
BC107057.2 | - | 9 | 701 | |||
BC148185.2 | - | 8 | 710 |