Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145548.1 | NP_001139020.1 | 9 | 1310 | ||
NM_017740.2 | NP_060210.2 | 8 | 1199 | |||
XM_017023434.1 | XP_016878923.1 | 6 | 735 | |||
XM_017023435.1 | XP_016878924.1 | 6 | 595 | |||
XM_017023436.1 | XP_016878925.1 | 6 | 710 | |||
GenBank mRNA | AK000286.1 | - | 8 | 1199 | ||
AK001654.1 | - | 6 | 585 | |||
AK225768.1 | - | 8 | 1151 | |||
BC017702.1 | - | 6 | 560 | 4394921 | ||
BC018772.1 | - | 9 | 1297 | 4869114 |