Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMBRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267782.1 | NP_001254711.1 | 10 | 2501 | ||
NM_001300731.1 | NP_001287660.1 | 8 | 2492 | |||
NM_017749.3 | NP_060219.2 | 9 | 2220 | |||
XM_005253009.3 | XP_005253066.1 | 8 | 2488 | |||
XM_005253014.3 | XP_005253071.1 | 9 | 2218 | |||
XM_006718259.2 | XP_006718322.1 | 8 | 2369 | |||
XM_017018008.1 | XP_016873497.1 | 8 | 2488 | |||
GenBank mRNA | AB051523.1 | - | 8 | 2444 | ||
AK000301.1 | - | 10 | 2451 | |||
AK023197.1 | - | 2 | 743 | |||
AK302808.1 | - | 8 | 2550 | |||
AK304530.1 | - | 5 | 919 | |||
BC045609.1 | - | 9 | 2220 | 5296472 |