Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FLVCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195283.1 | NP_001182212.1 | 9 | 970 | ||
NM_017791.2 | NP_060261.2 | 9 | 1852 | |||
GenBank mRNA | AF456126.1 | - | 9 | 1476 | ||
AK000378.1 | - | 9 | 1800 | |||
AK027804.1 | - | 8 | 740 | |||
AK297002.1 | - | 9 | 970 | |||
AY260572.1 | - | 9 | 1832 | |||
AY260577.1 | - | 9 | 1832 | |||
BC019087.2 | - | 9 | 1815 | 4866427 |