Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018000.2 | NP_060470.2 | 4 | 724 | ||
XM_011511468.2 | XP_011509770.1 | 5 | 724 | |||
GenBank mRNA | AF068290.1 | - | 3 | 223 | ||
AK000978.1 | - | 4 | 694 | |||
BC032747.1 | - | 4 | 733 | 5573149 | ||
BC082990.1 | - | 4 | 714 | 6159624 | ||
CX781801.1 | - | 4 | 719 |