Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018198.3 | NP_060668.2 | 11 | 1263 | ||
XM_017001745.1 | XP_016857234.1 | 10 | 1116 | |||
GenBank mRNA | AF306695.1 | - | 11 | 1177 | ||
AK001599.1 | - | 11 | 1172 | |||
AK095386.1 | - | 3 | 194 | |||
AK294539.1 | - | 10 | 1086 | |||
AK298378.1 | - | 11 | 1149 | |||
AL833841.1 | - | 11 | 1137 | |||
AY336749.1 | - | 11 | 1173 | |||
BC006086.1 | - | 11 | 1159 | 3946414 | ||
BC008772.1 | - | 11 | 1164 |