Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018198.3 | NP_060668.2 | 2 | 280 | ||
XM_017001745.1 | XP_016857234.1 | 2 | 207 | |||
GenBank mRNA | AF306694.1 | - | 2 | 179 | ||
AF306695.1 | - | 2 | 194 | |||
AK001599.1 | - | 2 | 189 | |||
AK294539.1 | - | 2 | 177 | |||
AK298378.1 | - | 2 | 166 | |||
AL109978.1 | - | 1 | 102 | |||
AL833841.1 | - | 2 | 154 | |||
AY336749.1 | - | 2 | 190 | |||
BC006086.1 | - | 2 | 176 | 3946414 | ||
BC008772.1 | - | 2 | 181 | |||
BC014145.2 | - | 2 | 188 | 4762069 | ||
DB461237.1 | - | 2 | 282 |