Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H2AFJ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177925.3 | NP_808760.1 | 1 | 232 | ||
NR_027716.2 | 1 | 232 | ||||
GenBank mRNA | AK001765.1 | - | 1 | 163 | ||
BC003602.1 | - | 1 | 153 | 3354610 | ||
BM689354.1 | - | 1 | 236 | |||
BU594879.1 | - | 1 | 210 | |||
DA808847.1 | - | 1 | 233 | |||
DB084616.1 | - | 1 | 233 |