Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146688.1 | NP_001140160.1 | 20 | 3181 | ||
NM_018433.5 | NP_060903.2 | 20 | 3450 | |||
XM_006712051.3 | XP_006712114.1 | 19 | 3148 | |||
XM_017004493.1 | XP_016859982.1 | 20 | 3469 | |||
XM_017004494.1 | XP_016859983.1 | 21 | 3292 | |||
GenBank mRNA | AB018285.2 | - | 20 | 3213 | ||
AK074314.1 | - | 6 | 847 | |||
AK304276.1 | - | 18 | 2967 | |||
AL832150.1 | - | 17 | 5129 | |||
BC038297.2 | - | 20 | 3227 | |||
BX640698.1 | - | 20 | 3251 | |||
BX647271.1 | - | 20 | 3219 | |||
BX647752.1 | - | 20 | 3279 | |||
BX648841.1 | - | 20 | 3182 | |||
BX649109.1 | - | 17 | 6083 | |||
CR749581.1 | - | 20 | 3218 |