Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VIMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018445.5 | NP_060915.2 | 5 | 530 | ||
NM_203472.2 | NP_982298.2 | 5 | 530 | |||
GenBank mRNA | AF157317.1 | - | 5 | 457 | ||
AF328864.1 | - | 5 | 457 | |||
AK026455.1 | - | 5 | 493 | |||
AK097011.1 | - | 3 | 1720 | |||
AK225955.1 | - | 5 | 456 | |||
AK314723.1 | - | 5 | 492 | |||
AY324824.1 | - | 5 | 457 | |||
AY618665.1 | - | 5 | 427 | |||
BC005840.2 | - | 5 | 453 | 2967406 | ||
BC107774.1 | - | 5 | 460 | 6450503 | ||
BG829960.1 | - | 5 | 530 |