Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRKCI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002740.5 | NP_002731.4 | 18 | 1964 | ||
XM_017006860.1 | XP_016862349.1 | 17 | 1871 | |||
GenBank mRNA | AB451443.1 | - | 18 | 1699 | ||
AK315342.1 | - | 18 | 1892 | |||
BC022016.2 | - | 18 | 1959 | 4823853 | ||
BC042405.1 | - | 6 | 730 | |||
BQ014217.1 | - | 1 | 523 | |||
L18964.1 | - | 18 | 1963 | |||
L33881.1 | - | 18 | 1903 |