Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018728.3 | NP_061198.2 | 19 | 2392 | ||
XM_011521781.2 | XP_011520083.1 | 19 | 2392 | |||
XM_017022408.1 | XP_016877897.1 | 18 | 2257 | |||
XM_017022409.1 | XP_016877898.1 | 14 | 1699 | |||
XM_017022410.1 | XP_016877899.1 | 18 | 2242 | |||
GenBank mRNA | AF272390.1 | - | 19 | 2249 | ||
AK304136.1 | - | 20 | 2507 | |||
BC144611.1 | - | 19 | 2177 |