Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC50A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122837.1 | NP_001116309.1 | 4 | 471 | ||
NM_001122839.1 | NP_001116311.1 | 4 | 433 | |||
NM_001287586.1 | NP_001274515.1 | 4 | 498 | |||
NM_001287587.1 | NP_001274516.1 | 5 | 615 | |||
NM_001287588.1 | NP_001274517.1 | 4 | 471 | |||
NM_001287589.1 | NP_001274518.1 | 3 | 309 | |||
NM_001287590.1 | NP_001274519.1 | 4 | 628 | |||
NM_001287591.1 | NP_001274520.1 | 5 | 652 | |||
NM_001287592.1 | NP_001274521.1 | 3 | 384 | |||
NM_018845.3 | NP_061333.2 | 5 | 595 | |||
XM_005245339.4 | XP_005245396.1 | 5 | 781 | |||
XM_005245340.3 | XP_005245397.1 | 5 | 862 | |||
XM_005245341.4 | XP_005245398.1 | 5 | 813 | |||
XM_005245342.3 | XP_005245399.1 | 4 | 421 | |||
XM_006711453.2 | XP_006711516.1 | 5 | 595 | |||
GenBank mRNA | AF126023.1 | - | 5 | 555 | ||
AF126024.1 | - | 4 | 420 | |||
AK289551.1 | - | 5 | 596 | |||
BC005943.1 | - | 5 | 539 | 4076256 | ||
BC009621.1 | - | 4 | 395 | 3896154 | ||
BC010869.1 | - | 4 | 386 | |||
BC012378.1 | - | 4 | 420 | |||
BE958344.1 | - | 452 | ||||
BM806886.1 | - | 5 | 639 | |||
BU160680.1 | - | 5 | 537 | |||
BU564973.1 | - | 4 | 458 | |||
BX093804.1 | - | 3 | 301 | |||
CR457148.1 | - | 5 | 495 | |||
X84712.1 | - | 3 | 180 |