Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC50A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122837.1 | NP_001116309.1 | 5 | 628 | ||
NM_001122839.1 | NP_001116311.1 | 5 | 590 | |||
NM_001287586.1 | NP_001274515.1 | 5 | 655 | |||
NM_001287587.1 | NP_001274516.1 | 6 | 772 | |||
NM_001287588.1 | NP_001274517.1 | 5 | 646 | |||
NM_001287589.1 | NP_001274518.1 | 4 | 466 | |||
NM_001287590.1 | NP_001274519.1 | 5 | 785 | |||
NM_001287591.1 | NP_001274520.1 | 6 | 809 | |||
NM_001287592.1 | NP_001274521.1 | 4 | 541 | |||
NM_018845.3 | NP_061333.2 | 6 | 752 | |||
XM_005245339.4 | XP_005245396.1 | 6 | 938 | |||
XM_005245340.3 | XP_005245397.1 | 6 | 1019 | |||
XM_005245341.4 | XP_005245398.1 | 6 | 970 | |||
XM_005245342.3 | XP_005245399.1 | 5 | 578 | |||
XM_006711453.2 | XP_006711516.1 | 6 | 770 | |||
GenBank mRNA | AF126023.1 | - | 6 | 712 | ||
AF126024.1 | - | 5 | 577 | |||
AK289551.1 | - | 6 | 753 | |||
BC005943.1 | - | 6 | 696 | 4076256 | ||
BC009621.1 | - | 5 | 552 | 3896154 | ||
BC010869.1 | - | 5 | 543 | |||
BC012378.1 | - | 5 | 577 | |||
BX093804.1 | - | 4 | 458 | |||
X84712.1 | - | 4 | 337 |