Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002751.6 | NP_002742.3 | 3, 4 | 403 | ||
NR_110887.1 | 3, 4 | 403 | ||||
GenBank mRNA | AF001008.1 | - | 3, 4 | 302 | ||
AF001174.1 | - | 3, 4 | 308 | |||
AF031135.1 | - | 3, 4 | 317 | |||
AK308969.1 | - | 3, 4 | 319 | |||
AK309046.1 | - | 3, 4 | 505 | |||
BC027933.1 | - | 3, 4 | 360 | 5199628 | ||
BM857051.1 | - | 3, 4 | 387 | |||
BX361670.2 | - | 3, 4 | 387 | |||
BX444192.2 | - | 3, 4 | 402 | |||
CR456514.1 | - | 3, 4 | 304 | |||
U53442.1 | - | 3, 4 | 321 | |||
U92268.1 | - | 3, 4 | 306 | |||
Y14440.1 | - | 3, 4 | 308 |