Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMA6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282232.1 | NP_001269161.1 | 4, 5 | 612 | ||
NM_001282233.1 | NP_001269162.1 | 3, 4 | 469 | |||
NM_001282234.1 | NP_001269163.1 | 4, 5 | 489 | |||
NM_002791.2 | NP_002782.1 | 4, 5 | 564 | |||
XM_017021468.1 | XP_016876957.1 | 4, 5 | 1900 | |||
GenBank mRNA | AK298920.1 | - | 3, 4 | 437 | ||
AK302008.1 | - | 4, 5 | 414 | |||
AK313011.1 | - | 4, 5 | 490 | |||
AK316223.1 | - | 4, 5 | 478 | |||
BC002979.1 | - | 4, 5 | 455 | 3542966 | ||
BC017882.1 | - | 4, 5 | 477 | 4278372 | ||
BC022354.1 | - | 4, 5 | 434 | 4292526 | ||
BC023659.1 | - | 4, 5 | 482 | 4869452 | ||
BC070137.1 | - | 4, 5 | 486 | 6458246 | ||
BG701535.1 | - | 4, 5 | 545 | |||
BQ009843.1 | - | 3, 4 | 524 | |||
BU657545.1 | - | 564 | ||||
CR456944.1 | - | 4, 5 | 404 | |||
X59417.1 | - | 4, 5 | 466 | |||
X61972.1 | - | 3, 4 | 329 |