Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020147.3 | NP_064532.1 | 3 | 885 | ||
GenBank mRNA | AK313472.1 | - | 3 | 883 | ||
AL360202.1 | - | 3 | 844 | |||
BC027857.1 | - | 3 | 866 | 5222191 | ||
BC072414.1 | - | 3 | 873 | 6167441 |