Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020153.3 | NP_064538.3 | 4 | 510 | ||
XM_011542905.2 | XP_011541207.1 | 4 | 3252 | |||
XM_017018017.1 | XP_016873506.1 | 3 | 249 | |||
GenBank mRNA | AK025480.1 | - | 4 | 469 | ||
DA251912.1 | - | 4 | 513 |