Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277061.1 | NP_001263990.1 | 9, 10 | 1346 | ||
NM_001277062.1 | NP_001263991.1 | 7, 8 | 1093 | |||
NM_001277064.1 | NP_001263993.1 | 6, 7 | 934 | |||
NM_001277068.1 | NP_001263997.1 | 6, 7 | 699 | |||
NM_020194.5 | NP_064579.3 | 8, 9 | 1083 | |||
XM_005246682.1 | XP_005246739.1 | 8, 9 | 1191 | |||
XM_005246683.1 | XP_005246740.1 | 8, 9 | 1088 | |||
XM_005246685.3 | XP_005246742.1 | 8, 9 | 1107 | |||
XM_005246687.3 | XP_005246744.1 | 7, 8 | 1032 | |||
XM_006712637.1 | XP_006712700.1 | 7, 8 | 990 | |||
XM_006712638.1 | XP_006712701.1 | 6, 7 | 915 | |||
XM_006712639.2 | XP_006712702.1 | 5, 6 | 756 | |||
XM_011511500.1 | XP_011509802.1 | 9, 10 | 1138 | |||
XM_011511501.1 | XP_011509803.1 | 8, 9 | 1159 | |||
XM_017004512.1 | XP_016860001.1 | 8, 9 | 1255 | |||
GenBank mRNA | AF246239.1 | - | 8, 9 | 1083 | ||
AF258660.1 | - | 9, 10 | 1201 | |||
AK026137.1 | - | 7, 8 | 994 | |||
AL517791.3 | - | 6, 7 | 809 | |||
AL833032.1 | - | 2, 3 | 723 | |||
BC000797.1 | - | 5, 6 | 606 | 3451868 | ||
DA737008.1 | - | 6, 7 | 673 |