Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITHD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020362.4 | NP_065095.2 | 3, 4 | 445 | ||
XM_011541804.1 | XP_011540106.1 | 2, 3 | 370 | |||
GenBank mRNA | AF218024.1 | - | 3, 4 | 426 | ||
AF221595.1 | - | 3, 4 | 437 | |||
AF271784.1 | - | 3, 4 | 441 | |||
AK296662.1 | - | 2, 3 | 370 | |||
AK310330.1 | - | 3, 4 | 392 | |||
AK313005.1 | - | 3, 4 | 413 | |||
BC017208.2 | - | 3, 4 | 406 | 3943360 |