Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270427.1 | NP_001257356.1 | 9 | 1402 | ||
NM_005047.3 | NP_005038.1 | 10 | 1531 | |||
XM_017014928.1 | XP_016870417.1 | 9 | 1331 | |||
GenBank mRNA | AK026925.1 | - | 6 | 834 | ||
AK291051.1 | - | 10 | 1432 | |||
AK303007.1 | - | 9 | 1332 | |||
BC014478.1 | - | 10 | 1420 | 4878087 | ||
D31889.1 | - | 10 | 1418 | |||
S79862.1 | - | 10 | 1422 |