Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSMD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002812.4 | NP_002803.2 | 1 | 334 | ||
GenBank mRNA | AK301771.1 | - | 1 | 345 | ||
AK304486.1 | - | 1 | 96 | |||
BC001164.1 | - | 1 | 85 | 3528096 | ||
BC065006.1 | - | 1 | 73 | 6055235 | ||
BI560993.1 | - | 1 | 318 | |||
BP369299.1 | - | 1 | 348 | |||
BQ684248.1 | - | 1 | 97 | |||
D38047.1 | - | 1 | 73 | |||
EU668354.1 | - | 1 | 334 |