Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYZL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199951.2 | NP_001186880.1 | 3 | 491 | ||
NM_020426.3 | NP_065159.1 | 3 | 605 | |||
GenBank mRNA | AY359018.1 | - | 2 | 409 | ||
AY742214.1 | - | 2 | 381 | |||
BC054481.1 | - | 3 | 451 | 6620264 | ||
BG195727.1 | - | 2 | 459 | |||
GQ472214.1 | - | 3 | 451 |