Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167882.1 | NP_001161354.1 | 3 | 1344 | ||
NM_020337.2 | NP_065070.1 | 4 | 2619 | |||
XM_017008471.1 | XP_016863960.1 | 3 | 3352 | |||
GenBank mRNA | AB033049.2 | - | 1 | 563 | ||
AK091516.1 | - | 1 | 635 | |||
AK127496.1 | - | 3 | 1470 | |||
AK291068.1 | - | 4 | 2619 | |||
AK295135.1 | - | 3 | 1344 | |||
BX640674.1 | - | 3 | 1136 |