Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGDR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000953.2 | NP_000944.1 | 2 | 1090 | ||
NM_001281469.1 | NP_001268398.1 | 3 | 1181 | |||
XM_005267891.3 | XP_005267948.1 | 2 | 1547 | |||
GenBank mRNA | BC040968.1 | - | 2 | 1090 | 5769056 | |
CD013871.1 | - | 3 | 1083 | |||
EF577397.1 | - | 2 | 988 | |||
U31099.1 | - | 2 | 344 |
Set Membership: |
G-protein coupled receptor |