Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CREBZF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039618.2 | NP_001034707.1 | 1 | 429 | ||
NR_028024.1 | 1 | 429 | ||||
NR_028025.1 | 1 | 429 | ||||
NR_028026.1 | 1 | 429 | ||||
NR_028027.1 | 1 | 429 | ||||
XM_006718642.3 | XP_006718705.1 | 1 | 439 | |||
XM_011545195.2 | XP_011543497.1 | 1 | 439 | |||
XM_017018086.1 | XP_016873575.1 | 1 | 439 | |||
XM_017018087.1 | XP_016873576.1 | 1 | 439 | |||
XM_017018088.1 | XP_016873577.1 | 1 | 439 | |||
XM_017018089.1 | XP_016873578.1 | 1 | 439 | |||
XM_017018090.1 | XP_016873579.1 | 1 | 439 | |||
XM_017018091.1 | XP_016873580.1 | 1 | 439 | |||
XM_017018092.1 | XP_016873581.1 | 1 | 439 | |||
GenBank mRNA | AF039942.1 | - | 1 | 377 | ||
BC060807.1 | - | 1 | 420 | 30345789 | ||
BF063403.1 | - | 1 | 481 | |||
DB212122.1 | - | 1 | 429 |