Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSTC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267817.1 | NP_001254746.1 | 1 | 83 | ||
NM_001267818.1 | NP_001254747.1 | 1 | 83 | |||
NM_021227.3 | NP_067050.1 | 1 | 83 | |||
GenBank mRNA | AF068297.1 | - | 1 | 46 | ||
AF161425.1 | - | 1 | 67 | |||
AF201937.1 | - | 1 | 74 | |||
AF343342.1 | - | 1 | 12 | |||
AK312185.1 | - | 1 | 27 | |||
AW081061.1 | - | 1 | 84 | |||
BC016321.1 | - | 1 | 71 | 4075646 | ||
BC054857.1 | - | 1 | 73 | 4093030 | ||
BM471860.1 | - | 11 | ||||
BM541430.1 | - | 1 | 34 | |||
BM546028.1 | - | 1 | 73 | |||
BQ435510.1 | - | 83 | ||||
CB110420.1 | - | 1 | 28 | |||
CN430159.1 | - | 1 | 21 |