Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006267.4 | NP_006258.3 | 20 | 5368 | ||
XM_005264002.2 | XP_005264059.1 | 20 | 5403 | |||
XM_005264003.2 | XP_005264060.1 | 20 | 5331 | |||
XM_005264004.2 | XP_005264061.1 | 20 | 5331 | |||
XM_005264005.4 | XP_005264062.1 | 20 | 5391 | |||
XM_011511575.2 | XP_011509877.1 | 20 | 5306 | |||
XM_011511576.2 | XP_011509878.1 | 21 | 5154 | |||
XM_017004623.1 | XP_016860112.1 | 20 | 5331 | |||
GenBank mRNA | AB209483.1 | - | 17 | 4984 | ||
D42063.1 | - | 20 | 5369 |