Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002887.3 | NP_002878.2 | 10 | 1206 | ||
GenBank mRNA | AK222797.1 | - | 10 | 1182 | ||
AK302161.1 | - | 9 | 1064 | |||
AK314795.1 | - | 10 | 1194 | |||
BC000528.2 | - | 10 | 1184 | 2961565 | ||
BC014619.2 | - | 10 | 1184 | 2961565 | ||
BT007394.1 | - | 10 | 1165 | |||
S80343.1 | - | 10 | 1192 |