Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RASGRF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006909.2 | NP_008840.1 | 18 | 2795 | ||
XM_005248565.1 | XP_005248622.1 | 18 | 2812 | |||
XM_017009682.1 | XP_016865171.1 | 18 | 2477 | |||
GenBank mRNA | AF023130.1 | - | 18 | 2745 | ||
AF181250.1 | - | 18 | 2745 | |||
BC126112.1 | - | 18 | 2795 | 8991828 | ||
BC136296.1 | - | 18 | 2795 | 9020283 | ||
BC143870.1 | - | 18 | 2795 |