Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193272.1 | NP_001180201.1 | 10 | 1200 | ||
NM_001193273.1 | NP_001180202.1 | 10 | 1144 | |||
NM_005057.3 | NP_005048.2 | 10 | 1200 | |||
XM_011509833.2 | XP_011508135.1 | 10 | 1198 | |||
GenBank mRNA | AK290137.1 | - | 10 | 1153 | ||
AK296978.1 | - | 10 | 1098 | |||
AK297540.1 | - | 9 | 1437 | |||
BC037284.2 | - | 10 | 1153 | 5266066 | ||
BC053856.1 | - | 10 | 1171 | 5576406 | ||
BC075059.2 | - | 10 | 1098 | 30915584 | ||
BC075060.2 | - | 10 | 1098 | 30915368 |