Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REG1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002909.4 | NP_002900.2 | 2 | 121 | ||
GenBank mRNA | AF172331.1 | - | 1 | 68 | ||
AK291981.1 | - | 2 | 89 | |||
AK307292.1 | - | 2 | 218 | |||
AK311087.1 | - | 2 | 89 | |||
BC005350.2 | - | 1 | 65 | 3950349 | ||
BC110438.2 | - | 2 | 84 | |||
BC118544.1 | - | 2 | 83 | |||
BC118621.1 | - | 2 | 83 | |||
BM769434.1 | - | 2 | 133 | |||
M18963.1 | - | 1 | 73 |