Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RFX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000635.3 | NP_000626.2 | 9, 10 | 1285 | ||
NM_134433.2 | NP_602309.1 | 8, 9 | 1210 | |||
XM_011528171.2 | XP_011526473.1 | 10, 11 | 1251 | |||
XM_011528172.1 | XP_011526474.1 | 9, 10 | 1108 | |||
XM_017027107.1 | XP_016882596.1 | 8, 9 | 1135 | |||
XM_017027108.1 | XP_016882597.1 | 7, 8 | 1060 | |||
GenBank mRNA | AK024288.1 | - | 6, 7 | 824 | ||
AK291196.1 | - | 8, 9 | 1089 | |||
AK302661.1 | - | 3, 4 | 273 | |||
AK303321.1 | - | 6, 7 | 594 | |||
BC028579.1 | - | 9, 10 | 1197 | 4829582 | ||
BC071571.1 | - | 9, 10 | 1166 | 5276369 | ||
BX648130.1 | - | 9, 10 | 1120 | |||
X76091.1 | - | 9, 10 | 1175 |