Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELAC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165962.1 | NP_001159434.1 | 7, 8 | 729 | ||
NM_018127.6 | NP_060597.4 | 8, 9 | 849 | |||
NM_173717.1 | NP_776065.1 | 8, 9 | 849 | |||
GenBank mRNA | AF304370.1 | - | 8, 9 | 732 | ||
AK001392.1 | - | 8, 9 | 806 | |||
AK094687.1 | - | 8, 9 | 733 | |||
AK125030.1 | - | 7, 8 | 994 | |||
AK298397.1 | - | 7, 8 | 664 | |||
AK300011.1 | - | 8, 9 | 724 | |||
AK304589.1 | - | 3, 4 | 206 | |||
BC001939.1 | - | 8, 9 | 792 | 2820640 | ||
BC004158.2 | - | 8, 9 | 792 | 2820640 | ||
CR457261.1 | - | 8, 9 | 732 | |||
KU178737.1 | - | 8, 9 | 732 |